pH Electrode Solutions, WTW
Supplier: YSI INC
109705 109648
80085-322EA 102.27 CAD
80085-322 16467-102 89207-612
pH Electrode Solutions, WTW
Electrolytes Electrode Filling Solutions
Solutions for operation, cleaning and storage of electrodes - simple refilling and safe storage warranted.
Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes. With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte.