Conductivity Meters, Handheld, Cond 3110 / 3310, WTW
Supplier: WTW
2CA100 2CA200 2CA201 2CA204 2CA300 2CA301
97043-140EA 2557.36 CAD
97043-140 97043-144 97043-146 97043-148 97043-150 97043-154 97043-156 97043-158 97043-160
Conductivity Meters, Handheld, Cond 3110 / 3310, WTW
Electrochemistry Meters Conductivity Meters
Rugged lines of conductivity meters with a range of features, benefits, and price points to meet every application and budget!
  • Rugged, waterproof housing is great for the field or the lab
  • Durable, I67-rated meters feature a 100% waterproof and easy-to-clean keypad
  • Silicone rubber keys provide tactile feedback for error-free operation
Cond 3110 - Designed for users who desire a simple-to-use, yet dependable instrument. The large graphic display allows for excellent readability even under difficult lighting conditions. A built-in calibration timerMore Product Information