Quartz Flat Bottom Flasks, Chemglass
Supplier: Chemglass
CG-1500-Q-14 CG-1500-Q-04 CG-1500-Q-18 CG-1500-Q-02 CG-1500-Q-08 CG-1500-Q-12
89028-506EA 447.67 CAD
89028-506 89028-516 89028-508 89028-518 89028-512 89028-522
Quartz Flat Bottom Flasks, Chemglass
Flasks Flat Bottom Flasks
Single neck flat bottom Quartz flask with Standard taper outer joint.