BioBox, 81-Place, for 3, 4 and 5 ml CryoClear™ Vials, Globe Scientific
Supplier: Globe Scientific
3042B 3042G 3042R 3042Y
10118-736PK 162.77 CAD
10118-736 10118-738 10118-740 10118-742
BioBox, 81-Place, for 3, 4 and 5 ml CryoClear™ Vials, Globe Scientific
Boxes Cryoboxes
Produced from polycarbonate (PC), these storage boxes are designed to withstand temperatures of −196 to +121°C
  • Polycarbonate (PC)
  • Transparent lid for convenient viewing of contents
  • The lid is printed with numbers for easy sample identification
  • Lid has two angled corners to ensure the correct alignment of the well numbers
  • Autoclavable at +121 °C, 15 psi (1 bar) for 20 minutes.
  • Fits in stainless steel storage racks
The lid is transparent for viewing contents and has a printed numerical grid for easy sample identification. Each box features two angledMore Product Information