BioBox, 25-Place for 1 and 2 ml CryoClear™ Vials, Globe Scientific
Supplier: Globe Scientific
3039B 3039G 3039R 3039Y
10118-720PK 189.99 CAD
10118-720 10118-722 10118-724 10118-726
BioBox, 25-Place for 1 and 2 ml CryoClear™ Vials, Globe Scientific
Boxes Cryoboxes
Produced from polycarbonate (PC), these storage boxes are designed to withstand temperatures of −196 to +121 °C
  • Polycarbonate (PC)
  • Transparent lid for convenient viewing of contents
  • The lid is printed with numbers for easy sample identification
  • Lid has two angled corners to ensure the correct alignment of the well numbers
  • Autoclavable at +121 °C, 15 psi (1 bar) for 20 minutes.
  • Fits in stainless steel storage racks
The lid is transparent for viewing contents and has a printed numerical grid for easy sample identification. Each box features two angled corners to guarantee proper alignment of the grid. A frosted writing area provides the perfect surface for smear-resistant marking.
The boxes fit in stainless steel freezer racks and have vent holes for drainage and rapid freezing
Ordering information: Holds 25 vials (5×5 format). Includes a free vial picker.