CryoClear™ Cryogenic Vials, Internal Threads, Globe Scientific
Supplier: Globe Scientific
3001 3002 3004 3005 3006 3008
10118-678CS 518.52 CAD
10118-678 10118-680 10118-682 10118-684 10118-686 10118-688 10118-690
CryoClear™ Cryogenic Vials, Internal Threads, Globe Scientific
Vials
Innovation to protect precious samples.
  • Non-pyrogenic, non-cytotoxic, non-mutagenic, heavy metals free, BSE/TSE free
  • RNase, DNase, ATP, and Human DNA free
  • Tamper evident packaging
  • Autoclavable
The best and most leak-resistant cryogenic vials available for the long-term storage of your precious samples at temperatures as low as –196 °C.
The vials feature an innovative screw cap that is co-molded with a layer of thermoplastic elastomer (TPE). This special process provides the leak-proof benefits of an O-ring, while eliminating the risk of contamination and sample loss associated with the use of separate O-rings.
Each vial is printed with a unique barcode that may be used for automated data collection, accurate sample inventory or to conceal the sample's identity.
Packaging: Packaged in tamper-evident bags of 50.
Caution: Not for use in the liquid phase of liquid nitrogen.