Borosilicate Glass Culture Tubes, Globe Scientific
Supplier: Globe Scientific
1503 1505 1510 1512 1515 1517 1519 1522
490015-322CS 103.21 CAD
490015-322 490015-324 490015-326 490015-328 490015-330 490015-332 76264-510 76264-512
Borosilicate Glass Culture Tubes, Globe Scientific
Tubes Culture Tubes
These tubes are made from superior quality borosilicate glass to reduce pH changes and contaminants that are potentially leached from soda lime glass tubes and are used for tissue culture and blood banks.
  • Excellent chemical resistance and durability
  • Features well-rounded bottoms and smooth fire-polished rims
TubeSaver™ packaging system helps prevent breakage and contamination during shipping and storage. Dimensional specifications are rigidly controlled for reliable, uniform results.
Please note: The capacity given below indicates the brim capacityMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable