PHCbi VIP® Series Cryogenic Chest Freezer, –150 °C, PHC Corporation
Supplier: PHC Corporation
PHCbi
MDFC2156VANCPA MDF-C2156VANC
CA10046-900EA 70611.07 CAD
CA10046-900 10046-900
PHCbi VIP® Series Cryogenic Chest Freezer, –150 °C, PHC Corporation
Freezers Cryogenic Freezers
The PHCbi VIP® PLUS –150 °C cryogenic freezer offers insulating properties that help to provide unsurpassed uniformity in the chamber from top to bottom, side to side, and front to back. The –150 °C cryogenic freezer offers air phase cryogenic storage without the need for liquid nitrogen as the primary cooling medium.
  • Unsurpassed uniformity
  • VIP® PLUS insulation
  • Easy temperature monitoring
  • 6 (3 leveling feet on front base)
Uniform temperature of –150 °C is maintained with VIP® PLUS insulation technology for stable, long-term preservation of cells and tissues.
PHCbi's VIP® PLUS cryogenic series demonstrates innovative control and monitoring. The temperature inside the freezer can be set and monitored easily by means of precise microprocessor temperature control with an LCD graphic display. The controller utilizes a platinum RTD sensor, which is extremely precise and durable.
The temperature inside the freezer can further be set and monitored by means of precise microprocessor temperature control with an LCD graphic display. The unit's mechanical refrigeration design is a better alternative to LN₂ storage as it eliminates concerns of cross-contamination and continual maintenance costs of LN₂ storage.
Certifications: QPS listed. RoHS compliant.