High-Capacity Double Mega Rack, Heathrow Scientific®
Supplier: Heathrow Scientific
HS120112 HS120113 HS120114 HS120115
89511-838EA 145.23 CAD
89511-838 89511-840 89511-842 89511-844
High-Capacity Double Mega Rack, Heathrow Scientific®
Racks Tube Racks
Fully assembled, polypropylene Mega Racks are great in clinical labs for storage of large groupings of samples
With a Mega Rack, you can place different test groupings into one rack and eliminate the need for multiple racks. They are made from sturdy, lightweight material for easy transport and include an alpha-numeric grid for quick location and retrieval of tubes.