DuraPorter™ Transport Box, Heathrow Scientific®
Supplier: VWR International
89511-782 89511-802 89511-804
89511-782CS 2842.33 CAD
89511-782 89511-802 89511-804
DuraPorter™ Transport Box, Heathrow Scientific®
Boxes Transport Boxes
Ideal for transporting biological and clinical samples, as well as instruments and products that need to be kept clean and dry under testing environmental conditions
The clarity of the DuraPorter™ box enables users to visibly check sample integrity, so that it can be opened using the right precautions. All boxes feature leak-proof, polycarbonate construction. Lids open a full 180°.