Beaker Cover, PTFE, Dynalon
Supplier: Dynalon
315304 315314 315374
13914-620EA 25.25 CAD
13914-620 13914-622 13914-634
Beaker Cover, PTFE, Dynalon
Beakers
Isostatically molded beaker covers are manufactured from virgin PTFE and machine finished to give a smooth surface for easy cleaning and contamination elimination.
Virtually immune to chemical attack. Working temperature range is from cryogenic to 260 °C.