Stir Bar, Color-Coded, PTFE, Dynalon
Supplier: Dynalon
304415-0003 304425-0003 304425-0005 304425-0004 304425-0007 304405-0005 304405-0004 304405-0007
30620-404CS 211.89 CAD
30620-404 30620-464 30620-466 30620-468 30620-470 30620-492 30620-494 30620-496 30620-498
Stir Bar, Color-Coded, PTFE, Dynalon
Stirrers Magnetic Stirring Bars
Color-coded for identification, these spin bars feature powerful permanent Alnico V magnetic cores
PTFE offers excellent chemical resistance. Always store in pairs to maintain magnetic strength. May leach colorant in very harsh chemicals.
Order Now
SPECIFICATIONS
Environmentally Preferable