Stir Bar, Round, PTFE, Dynalon
Supplier: Dynalon
303575 303705
58947-153CS 424.81 CAD
58947-153 58947-110
Stir Bar, Round, PTFE, Dynalon
Stirrers Magnetic Stirring Bars
Round stirring bars feature powerful permanent Alnico V magnetic cores
PTFE offers excellent chemical resistance and is autoclavable. Always store in pairs to maintain magnetic strength.