Stirring Rod, Solid, Dynalon
Supplier: Dynalon
303334-0003 303334-0004 303334-0000 303334-0001 303334-0002
30620-504CS 283.15 CAD
30620-504 30620-506 30620-508 30620-510 30620-512
Stirring Rod, Solid, Dynalon
Stirrers Stirring Rods
Pure PTFE solid stirring rod with flat tapered ends
Inert and will not scratch, the rod can be used up to 280°C. PTFE has excellent chemical resistance and is autoclavable.