Stir Bar, Cross Head, PTFE, Dynalon
Supplier: Dynalon
304495-0001 304495-0003 304495-0004
30620-406CS 333.06 CAD
30620-406 30620-408 30620-410
Stir Bar, Cross Head, PTFE, Dynalon
Stirrers Magnetic Stirring Bars
Designed for round or flat bottom labware and test tubes
Greater mixing action at bottom, side, and top of container, even at low stirring speeds. Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance and is autoclavable. Always store in pairs to maintain magnetic strength.