Stir Bar, Octagonal, PTFE, Dynalon
Supplier: Dynalon
303275-0004 303275-0011 303275-0005 303275-0002 303275-0003 303275-0001 303275-0006 303275-0007 303275-0016 303275-0008 303275-0014 303275-0010 303275-0019 303275-0009 303275-0018 303275-0021
58948-219EA 12.64 CAD
58948-219 58947-010 58949-127 30620-424 30620-426 30620-430 30620-434 30620-438 30620-440 30620-442 30620-446 30620-448 30620-450 30620-452 30620-454 30620-456
Stir Bar, Octagonal, PTFE, Dynalon
Stirrers Magnetic Stirring Bars
Octagonal stirring bars feature powerful permanent Alnico V magnetic cores
PTFE offers excellent chemical resistance. Always store in pairs to maintain magnetic strength.