Anti-RRM2B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-B67 600-401-B67S
RL600-401-B67EA 834.24 CAD
RL600-401-B67 RL600-401-B67S
Anti-RRM2B Rabbit Polyclonal Antibody
Antibodies
RRM2B/p53R2 antibody is suitable for IP, IHC, IF, ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 41 kDa in size corresponding to RRM2B1 by western blotting.

Recommended Dilutions: ELISA: 1:3000; Western Blot: 1 µg/ml;

Type: Primary
Antigen: RRM2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR