Anti-CASZ1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-B62 600-401-B62S
RL600-401-B62EA 834.24 CAD
RL600-401-B62 RL600-401-B62S
Anti-CASZ1 Rabbit Polyclonal Antibody
Antibodies
This antibody has been tested for use in ELISA and western blotting. This antiserum detects endogenous Casz1 Proteins, both the hCasz5 and hCasz11 isoforms. Expect a band approximately 125 kDa for hCasz5 and 190 kDa for hCaz11.

Recommended Dilutions: ELISA: 1:45000; Western Blot: 1:1000 - 1:5000;

Type: Primary
Antigen: CASZ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Drosophila, Chimpanzee, Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR