Anti-TRAF2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-B27 600-401-B27S
RL600-401-B27EA 834.24 CAD
RL600-401-B27 RL600-401-B27S
Anti-TRAF2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:60,000 - 1:250000; Western Blot: 1 µg/ml;

Type: Primary
Antigen: TRAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR