Anti-SAE1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-B24 600-401-B24S
RL600-401-B24EA 834.24 CAD
RL600-401-B24 RL600-401-B24S
Anti-SAE1 Rabbit Polyclonal Antibody
Antibodies
This purified antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~37 kDa in size corresponding to phosphorylated SAE1 Protein by western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 2µg/ml;

Type: Primary
Antigen: SAE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovine, Mouse, Rat, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR