Anti-IL33 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
209-401-B50 209-401-B50S
RL209-401-B50EA 834.24 CAD
RL209-401-B50 RL209-401-B50S
Anti-IL33 Rabbit Polyclonal Antibody
Antibodies
This purified antibody has been tested for use in neutralizations, ELISA and western blotting. Reactivity is also expected in radioimmunoassay.

Recommended Dilutions:

Type: Primary
Antigen: IL33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR