Anti-ALPI Sheep Polyclonal Antibody

Supplier: Rockland Immunochemical
200-6135-0100 200-6135S
CARL2006135100EA 834 CAD
CARL2006135100 CARL2006135S
Anti-ALPI Sheep Polyclonal Antibody
Antibodies
ALKALINE PHOSPHATASE antibody has been assayed against 1.0 ug of Alkaline Phosphatase [Calf Intestine] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Sheep IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000; Other Dilution: User Optimized

Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR