Anti-SOD1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4191-0100 200-4191S
CARL2004191100EA 834 CAD
CARL2004191100 CARL2004191S
Anti-SOD1 Rabbit Polyclonal Antibody
Antibodies
SUPEROXIDE DISMUTASE (SOD) Antibody has been assayed against 1.0 ug of Superoxide Dismutase [Bovine erythrocytes] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500 - 1:2000

Type: Primary
Antigen: SOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR