Anti-PRSS2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4180-0100 200-4180S
CARL2004180100EA 834 CAD
CARL2004180100 CARL2004180S
Anti-PRSS2 Rabbit Polyclonal Antibody
Antibodies
TRYPSINOGEN Antibody has been assayed against 1.0 ug of Trypsinogen [Bovine Pancreas] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000

Type: Primary
Antigen: PRSS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR