Anti-GALM Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4162-0100 200-4162S
CARL2004162100EA 834 CAD
CARL2004162100 CARL2004162S
Anti-GALM Rabbit Polyclonal Antibody
Antibodies
MUTAROTASE Antibody has been assayed against 1.0 ug of Mutarotase [Porcine Kidney] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100

Type: Primary
Antigen: GALM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR