Anti-DNASE1, DNL1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4161-0100 200-4161S
CARL2004161100EA 834 CAD
CARL2004161100 CARL2004161S
Anti-DNASE1, DNL1 Rabbit Polyclonal Antibody
Antibodies
Anti-DEOXYRIBONUCLEASE I has been assayed against 1.0 ug of Deoxyribonuclease A [Bovine Pancreas] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100

Type: Primary
Antigen: DNASE1, DNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR