Anti-CA2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4157-0100 200-4157S
CARL2004157100EA 834 CAD
CARL2004157100 CARL2004157S
Anti-CA2 Rabbit Polyclonal Antibody
Antibodies
Anti Carbonic Anhydrase II Antibody has been assayed against 1.0 µg of Carbonic Anhydrase II [Bovine Erythrocytes] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and TMBE.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000

Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR