Anti-AAO Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4148-0100 200-4148S
CARL2004148100EA 834 CAD
CARL2004148100 CARL2004148S
Anti-AAO Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:25000 - 1:125000; Western Blot: 1:2000 - 1:10000; Immunoprecipitation: 1:100

Type: Primary
Antigen: AAO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: cucurbita
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR