Anti-BMY1, AMYB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4147-0100 200-4147S
CARL2004147100EA 834 CAD
CARL2004147100 CARL2004147S
Anti-BMY1, AMYB Rabbit Polyclonal Antibody
Antibodies
BETA AMYLASE antibody has been assayed against 1.0 ug of b Amylase [Sweet Potato] in a standard sandwich ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Type: Primary
Antigen: BMY1, AMYB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: sweet potato
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR