Anti-ADH Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4143-0100 200-4143S
CARL2004143100EA 834 CAD
CARL2004143100 CARL2004143S
Anti-ADH Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100

Type: Primary
Antigen: ADH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Saccharomyces cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR