Anti-GOX Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-4137-0100 200-4137S
CARL2004137100EA 834 CAD
CARL2004137100 CARL2004137S
Anti-GOX Rabbit Polyclonal Antibody
Antibodies
Anti-Glucose Oxidase Antibody is suitable for western blotting, Immunochemistryand for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:35000 - 1:160000; Western Blot: 1:3500 - 1:16000; Immunochemistry: 1:2000 - 1:8000

Type: Primary
Antigen: gox
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Aspergillus niger
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR