Anti-CCL2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-B60 200-401-B60S
RL200-401-B60EA 834.24 CAD
RL200-401-B60 RL200-401-B60S
Anti-CCL2 Rabbit Polyclonal Antibody
Antibodies
This Protein-A purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 8.8 kDa in size corresponding to bovine CCL2 Protein by western blotting.

Recommended Dilutions: ELISA: 1:4500; Western Blot: 1 µg/ml;

Type: Primary
Antigen: CCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig, Human, Cat, Orangutan, Dog, Mouse, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR