Anti-GPC1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-B44 200-401-B44S
RL200-401-B44EA 834.24 CAD
RL200-401-B44 RL200-401-B44S
Anti-GPC1 Rabbit Polyclonal Antibody
Antibodies
Anti-Glypican-1 Protein A purified antibody has been tested for use in ELISA, and immunochemistry, and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~61 kDa in size.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunochemistry: 10µg/ml;

Type: Primary
Antigen: GPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR