Anti-HRPC Mouse Polyclonal Antibody

Supplier: Rockland Immunochemical
200-3138-0100 200-3138S
CARL2003138100EA 749 CAD
CARL2003138100 CARL2003138S
Anti-HRPC Mouse Polyclonal Antibody
Antibodies
Anti-Peroxidase (Horseradish) (Mouse) antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation, conjugation and most immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:3000; Immunochemistry: User Optimized; Other Dilution: User Optimized

Type: Tag
Antigen: HRPC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR