Anti-SHANK2 Mouse Monoclonal Antibody [clone: S23b-49]

Supplier: Rockland Immunochemical
200-301-G42
RL200-301-G42EA 854.78 CAD
RL200-301-G42
Anti-SHANK2 Mouse Monoclonal Antibody [clone: S23b-49]
Antibodies
Anti-SHANK Antibody is suitable for use in WB, Immunochemistryand IP. Expect a band approximately ~160kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions:

Type: Primary
Antigen: SHANK2
Clonality: Monoclonal
Clone: S23b-49
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR