Anti-RHO Mouse Monoclonal Antibody [clone: 1D4]

Supplier: Rockland Immunochemical
200-301-G37
RL200-301-G37EA 854.78 CAD
RL200-301-G37
Anti-RHO Mouse Monoclonal Antibody [clone: 1D4]
Antibodies
Anti-Rhodopsin Antibody is recommended for use in Immunofluorescence microscopy, IHC, IP, WB, and ELISA. Expect a band approximately ~40kDa corresponding to specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:1000;

Type: Primary
Antigen: RHO
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Vertebrates, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR