Anti-KCNAB2 Mouse Monoclonal Antibody [clone: S17-70]

Supplier: Rockland Immunochemicals
200-301-G08
RL200-301-G08EA 812.12 CAD
RL200-301-G08
Anti-KCNAB2 Mouse Monoclonal Antibody [clone: S17-70]
Antibodies
Anti-KvBeta2 K+ channel Antibody is suitable for use in WB, IP, and IHC. Expect a band approximately ~39kDa(major), ~41kDa (Minor) on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions:

Type: Primary
Antigen: KCNAB2
Clonality: Monoclonal
Clone: S17-70
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human, Zebrafish, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR