Anti-TRPC6 Mouse Monoclonal Antibody [clone: 3F2.H10.F2]

Supplier: Rockland Immunochemical
200-301-B59 200-301-B59S
RL200-301-B59EA 854.78 CAD
RL200-301-B59 RL200-301-B59S
Anti-TRPC6 Mouse Monoclonal Antibody [clone: 3F2.H10.F2]
Antibodies
This monoclonal antibody is suitable for ELISA, immunochemistry and western blotting. Expect a band approximately 106 kDa in size corresponding to TRPC6 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunochemistry: 2.5 µg/ml;

Type: Primary
Antigen: TRPC6
Clonality: Monoclonal
Clone: 3F2.H10.F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR