Anti-zwf Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-1153-0100 200-1153S
CARL2001153100EA 834 CAD
CARL2001153100 CARL2001153S
Anti-zwf Goat Polyclonal Antibody
Antibodies
GLUCOSE-6-PHOSPHATE DEHYDROGENASE Antibody has been assayed against 1.0 ug of Glucose-6-Phosphate Dehydrogenase [Leuconostoc mesenteroides] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG [H&L] (Rabbit) and (

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000; Other Dilution: User Optimized

Type: Primary
Antigen: zwf
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Leuconostoc mesenteroides
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR