Anti-BAG6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-B63 100-401-B63S
RL100-401-B63EA 834.24 CAD
RL100-401-B63 RL100-401-B63S
Anti-BAG6 Rabbit Polyclonal Antibody
Antibodies
This Scythe polyclonal antibody has been tested for use in western blotting and is specific for mouse Scythe/Bat3 and its isoforms. Specific conditions for reactivity should be optimized by the end user. Expect multiple bands.

Recommended Dilutions: Western Blot: 1:5000 to 1:8000

Type: Primary
Antigen: BAG6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR