Anti-CD34 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 561]

Supplier: Biolegend
343610 343609
CA343610-BLEA 641.51 CAD
CA343610-BL CA343609-BL
Anti-CD34 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 561]
Antibodies
Brilliant Violet 421™ anti-human CD34 [561]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 561
Conjugation: Brilliant Violet® 421
Epitope:
Host:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR