Anti-CD160 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BY55]

Supplier: Biolegend
341207 341208
CA341207-BLEA 256.92 CAD
CA341207-BL CA341208-BL
Anti-CD160 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BY55]
Antibodies
APC anti-human CD160 [BY55]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD160
Clonality: Monoclonal
Clone: BY55
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR