Anti-CD161 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HP-3G10]

Supplier: Biolegend
339914 339913
CA339914-BLEA 649.49 CAD
CA339914-BL CA339913-BL
Anti-CD161 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HP-3G10]
Antibodies
Brilliant Violet 421™ anti-human CD161 [HP-3G10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: Brilliant Violet® 421
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR