Anti-CD8a Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: RPA-T8]

Supplier: Biolegend
301047 301048
CA301047-BLEA 280.86 CAD
CA301047-BL CA10757-504
Anti-CD8a Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: RPA-T8]
Antibodies
Brilliant Violet 510™ anti-human CD8a [RPA-T8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Pigtailed Macaque, Sooty Mangabey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: RPA-T8
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus;Pigtailed Macaque;Sooty Mangabey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR