Anti-CD283 (TLR3) Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11F8]

Supplier: Biolegend
141905 141906
CA141905-BLEA 284.05 CAD
CA141905-BL CA141906-BL
Anti-CD283 (TLR3) Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11F8]
Antibodies
APC anti-mouse CD283 (TLR3) [11F8]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD283 (TLR3)
Clonality: Monoclonal
Clone: 11F8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR