Anti-CD27 Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: LG.3A10]

Supplier: Biolegend
124217 124218
CA124217-BLEA 178.73 CAD
CA124217-BL CA124218-BL
Anti-CD27 Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: LG.3A10]
Antibodies
Pacific Blue™ anti-mouse/rat/human CD27 [LG.3A10]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat, Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: LG.3A10
Conjugation: PB (Pacific Blue)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR