Anti-CD3 Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: 17A2]
Supplier: Biolegend
100233 100234
CA100233-BLEA 280.86 CAD
CA100233-BL CA10759-016
Anti-CD3 Rat Monoclonal Antibody (Brilliant Violet® 510) [clone: 17A2]
Antibodies
Brilliant Violet 510™ anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For flow cytometric staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.