Uridine

Supplier: HIMEDIA LABORATORIES PVT. CA

TC181-5G TC181-25G TC181-50G
CA89130-786EA 606.02 CAD
CA89130-786 CA89130-788 CA89130-790
Uridine
Uridine
Formula: C₉H₁₂N₂O₆
MDL Number: MFCD00006526
CAS Number: 58-96-8

Order Now

Specification Test Results

Molecular Weight 244.2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR