Guanidinium hydrochloride for molecular biology, Millipore®

Supplier: MilliporeSigma

Synonyms: Guanidinium chlorideGuanidine hydrochlorideGuanidine chlorideAminoformamidine hydrochlorideGuanadine hydrochlorideAminomethanamidine hydrochlorideGuanidine HCI

369080-1KG 369080-100GM 369080-1KG 369080-100GM
CA80502-626EA 429.63 CAD
80502-626 80502-628 CA80502-626 CA80502-628
Guanidinium hydrochloride for molecular biology, Millipore®
Guanidine hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
MDL Number: MFCD00013026
CAS Number: 50-01-1
https://ca.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg

Order Now

Specification Test Results

Molecular Weight 95.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR