BRAND® Pipette Tips, 200 µl, BrandTech®
Supplier: Brandtech
732028 732108 732208 732228 732008
10033-978PK 455.94 CAD
10033-978 10033-990 10034-002 10034-022 10034-110 10034-132
BRAND® Pipette Tips, 200 µl, BrandTech®
Pipette Tips
BRAND 200 µl pipette tips are a thin-walled low waste tip that fits virtually all pipettes with a yellow color code.
  • Fits most major brands of pipettes
  • Made under cleanroom conditions
  • All boxed/refill tips are DNA-/Rnase-/ATP-/endotoxin-free
All tips are produced under cleanroom conditions. Bulk tips are packed in resealable bags. New environmentally friendly refillable TipBox design is rigid and allows partial loading of multichannels. Hinged lid can be rotated 180° to be a lift-off design. Environmentally friendly Tip Refills reload empty TipBoxes,More Product Information